4-(2-Iodacetamido)-2,2,6,6-tetramethylpiperidin-1-oxyl (4-(2-Iodacetamido)-TEMPO freies Radikal), Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4-(2-Iodoacetamido)-2,2,6,6-tetramethylpiperidine 1-Oxyl, 4-(2-Iodoacetamido)-TEMPO

253367-10MG
SIAL253367-10MGEA 102 EUR
SIAL253367-10MG
4-(2-Iodacetamido)-2,2,6,6-tetramethylpiperidin-1-oxyl (4-(2-Iodacetamido)-TEMPO freies Radikal), Sigma-Aldrich®
4-(2-Iodacetamido)-2,2,6,6-tetramethylpiperidin-1-oxyl (4-(2-Iodacetamido)-TEMPO freies Radikal)
Formel: C₁₁H₂₀IN₂O₂
Molecular Weight: 339,2 g/mol
MDL: MFCD00010171
CAS-Nummer: 25713-24-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR