2-Ethylbenzoesäure, Sigma-Aldrich®

Lieferant: Merck


MQ100

253804-5G
SIAL253804-5GEA 171 EUR
SIAL253804-5G
2-Ethylbenzoesäure, Sigma-Aldrich®
2-Ethylbenzoic acid
Formel: C₉H₁₀O₂
Molecular Weight: 150.1700 g/mol
MDL: MFCD00045838
CAS-Nummer: 612-19-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR