2,3,4-Trihydroxybenzoesäure, Sigma-Aldrich®

Lieferant: Merck
Achtung


M-Clarity™ Qualitätsstufe = MQ100

253847-5G
SIAL253847-5GEA 83.4 EUR
SIAL253847-5G
2,3,4-Trihydroxybenzoesäure, Sigma-Aldrich®
2,3,4-Trihydroxybenzoic acid
Formel: C₇H₆O₅
Molecular Weight: 170,12 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00002447
CAS-Nummer: 610-02-6
EINECS: 210-205-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR