(Triphenylphosphin)gold(I)chlorid, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

254037-500MG 254037-5G
SIAL254037-500MGEA 192 EUR
SIAL254037-500MG SIAL254037-5G
(Triphenylphosphin)gold(I)chlorid, Sigma-Aldrich®
(Triphenylphosphine)gold(I) chloride
Formel: C₁₈H₁₅AuClP
MDL: MFCD00009588
CAS-Nummer: 14243-64-2
EINECS: 238-117-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR