Lithiumniobat, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Lithium niobium oxide
M-Clarity™ Qualitätsstufe = MQ100
254290-10G
SIAL254290-10GEA
53
EUR
SIAL254290-10G
Lithiumniobat, Sigma-Aldrich®
Lithiumniobat
Formel:
LiNbO₃ Molecular Weight: 147,85 g/mol Schmelzpunkt: 1257 °C Dichte: 4,638 g/cm³ (20 °C) |
MDL:
MFCD00011080 CAS-Nummer: 12031-63-9 EINECS: 234-755-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...