Zink(II)tellurid, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ100
254320-25G
SIAL254320-25GEA
116
EUR
SIAL254320-25G
Zink(II)tellurid, Sigma-Aldrich®
Zink(II)tellurid
Formel:
TeZn Molecular Weight: 192,99 g/mol Schmelzpunkt: 1294,85 °C Dichte: ∼6,3 g/cm³ (20 °C) |
MDL:
MFCD00043079 CAS-Nummer: 1315-11-3 EINECS: 215-260-2 Merck Index: 12,10297 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...