1,3,5-Trimethylhexahydro-1,3,5-triazin, Sigma-Aldrich®

Lieferant: Merck

254673-25G
SIAL254673-25GEA 105 EUR
SIAL254673-25G
1,3,5-Trimethylhexahydro-1,3,5-triazin, Sigma-Aldrich®
1,3,5-Trimethylhexahydro-1,3,5-triazin
Formel: C₆H₁₅N₃
MDL: MFCD00006057
CAS-Nummer: 108-74-7
EINECS: 203-612-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR