Tris(methylthio)methan, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

255068-5G
SIAL255068-5GEA 74.5 EUR
SIAL255068-5G
Tris(methylthio)methan, Sigma-Aldrich®
Tris(methylthio)methan
Formel: C₄H₁₀S₃
Molecular Weight: 154,32 g/mol
CAS-Nummer: 5418-86-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR