3-Aminobenzophenon, Sigma-Aldrich®

Lieferant: Merck

Synonyme: m-Aminobenzophenon

M-Clarity™ Qualitätsstufe = MQ100

255254-5G
SIAL255254-5GEA 71.2 EUR
SIAL255254-5G
3-Aminobenzophenon, Sigma-Aldrich®
3-Aminobenzophenon
Formel: C₁₃H₁₁NO
Molecular Weight: 197,24 g/mol
MDL: MFCD00017103
CAS-Nummer: 2835-78-1
EINECS: 220-614-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR