3-Brombiphenyl (PBB Nr. 2), Sigma-Aldrich®
Lieferant: Merck
Synonyme:
1-Brom-3-phenylbenzol
M-Clarity™ Qualitätsstufe = MQ100
255386-5G
SIAL255386-5GEA
54
EUR
SIAL255386-5G
3-Brombiphenyl (PBB Nr. 2), Sigma-Aldrich®
3-Brombiphenyl (PBB Nr. 2)
Formel:
C₁₂H₉Br Molecular Weight: 233.11 g/mol Siedepunkt: 299 Schmelzpunkt: 9 Dichte: 1.41 g/cm³ (20 °C) |
MDL:
MFCD00000082 CAS-Nummer: 2113-57-7 EINECS: 218-304-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...