1-Amino-4-methylpiperazin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4-Methylpiperazin-1-amine

M-Clarity™ Qualitätsstufe = MQ100

255688-25G
SIAL255688-25GEA 65.1 EUR
SIAL255688-25G
1-Amino-4-methylpiperazin, Sigma-Aldrich®
1-Amino-4-methylpiperazin
Formel: C₅H₁₃N₃
Molecular Weight: 115,18 g/mol
MDL: MFCD00006154
CAS-Nummer: 6928-85-4
EINECS: 230-053-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR