4-Chlor-2-nitrophenol, Sigma-Aldrich®
Lieferant: Merck
25580-250G
SIAL25580-250GEA
168
EUR
SIAL25580-250G
4-Chlor-2-nitrophenol, Sigma-Aldrich®
4-Chlor-2-nitrophenol
Formel:
C₆H₄ClNO₃ Molecular Weight: 173,56 g/mol Schmelzpunkt: 85…88 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007113 CAS-Nummer: 89-64-5 EINECS: 201-927-5 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...