(1S,2S)-1,2-Diphenyl-1,2-ethandiol, Sigma-Aldrich®

Lieferant: Merck

256269-25G 256269-5G
SIAL256269-25GEA 271 EUR
SIAL256269-25G SIAL256269-5G
(1S,2S)-1,2-Diphenyl-1,2-ethandiol, Sigma-Aldrich®
(S,S)-(-)-Hydrobenzoin
Formel: C₁₄H₁₄O₂
MDL: MFCD00064255
CAS-Nummer: 2325-10-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR