Chloroform : isoamyl alcohol (49:1) (v/v), Sigma-Aldrich®

Lieferant: Merck
Achtung


M-Clarity™ Qualitätsstufe = MQ200

25668-100ML 25668-500ML
SIAL25668-100MLEA 84.6 EUR
SIAL25668-100ML SIAL25668-500ML
Chloroform : isoamyl alcohol (49:1) (v/v), Sigma-Aldrich®
Chloroform : isoamyl alcohol
Formel: CHCl₃,C₅H₁₂O
UN: 1888
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR