Methylmagnesiumbromid 3 mol/l in Diethylether, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

257087-200ML
SIAL257087-200MLEA 226 EUR
SIAL257087-200ML
Methylmagnesiumbromid 3 mol/l in Diethylether, Sigma-Aldrich®
Methylmagnesium bromide
Formel: CH₃BrMg
Molecular Weight: 119,24 g/mol
MDL: MFCD00000041
CAS-Nummer: 75-16-1
EINECS: 200-844-1
UN: 1928
ADR: 4.3,I

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR