Phenylmagnesiumchlorid 2 mol/l in THF, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

257125-200ML
SIAL257125-200MLEA 176 EUR
SIAL257125-200ML
Phenylmagnesiumchlorid 2 mol/l in THF, Sigma-Aldrich®
Phenylmagnesium chloride
Formel: C₆H₅ClMg
Molecular Weight: 136,86 g/mol
MDL: MFCD00000464
CAS-Nummer: 100-59-4
EINECS: 202-868-8
UN: 3399
ADR: 4.3,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR