1H,1H-Pentadecafluor-1-octanol, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ200

257451-25G
SIAL257451-25GEA 154 EUR
SIAL257451-25G
1H,1H-Pentadecafluor-1-octanol, Sigma-Aldrich®
1H,1H-Pentadecafluor-1-octanol
Formel: C₈H₃F₁₅O
MDL: MFCD00004675
CAS-Nummer: 307-30-2
EINECS: 206-197-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR