1-Iodperfluordecan, Sigma-Aldrich®

Lieferant: Merck
Achtung

Synonyme: Perfluordecyliodid

M-Clarity™ Qualitätsstufe = MQ100

257842-5G
SIAL257842-5GEA 0 EUR
SIAL257842-5G
1-Iodperfluordecan, Sigma-Aldrich®
1-Iodperfluordecan
Formel: C₁₀F₂₁I
Molecular Weight: 645.98 g/mol
MDL: MFCD00001065
CAS-Nummer: 423-62-1
EINECS: 207-030-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR