3,6-Dibromcarbazol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3,6-Dibromo-9H-carbazole

M-Clarity™ Qualitätsstufe = MQ100

259004-5G
SIAL259004-5GEA 56.8 EUR
SIAL259004-5G
3,6-Dibromcarbazol, Sigma-Aldrich®
3,6-Dibromocarbazole
Formel: C₁₂H₇Br₂N
Molecular Weight: 325 g/mol
MDL: MFCD00004961
CAS-Nummer: 6825-20-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR