4-Acetoxyindol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4-Indolyl acetate

M-Clarity™ Qualitätsstufe = MQ100

259047-250MG
SIAL259047-250MGEA 61.6 EUR
SIAL259047-250MG
4-Acetoxyindol, Sigma-Aldrich®
4-Acetoxyindol
Formel: C₁₀H₉NO₂
Molecular Weight: 175,19 g/mol
MDL: MFCD00010678
CAS-Nummer: 5585-96-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR