Iron(III) meso-tetraphenylporphine chloride ≥94% (durch HPLC), Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

259071-500MG
SIAL259071-500MGEA 198 EUR
SIAL259071-500MG
Iron(III) meso-tetraphenylporphine chloride ≥94% (durch HPLC), Sigma-Aldrich®
Iron(III) meso-tetraphenylporphine chloride
Formel: C₄₄H₂₈ClFeN₄
Dichte: 1,276 g/cm³ (20 °C)
MDL: MFCD00012152
CAS-Nummer: 16456-81-8
EINECS: 213-025-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR