1,4,8,11-Tetraazacyclopentadecan, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1,4,8,12-Tetraazacyclopentadecane

M-Clarity™ Qualitätsstufe = MQ100

259152-1G
SIAL259152-1GEA 264 EUR
SIAL259152-1G
1,4,8,11-Tetraazacyclopentadecan, Sigma-Aldrich®
1,4,8,11-Tetraazacyclopentadecan
Formel: C₁₁H₂₆N₄
Molecular Weight: 214,35 g/mol
MDL: MFCD00010049
CAS-Nummer: 15439-16-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR