4-Brombenzhydrazid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4-Brombenzoesäurehydrazid, 4-Brombenzohydrazid, 4-Brombenzol-1-carbohydrazid

M-Clarity™ Qualitätsstufe = MQ200

259624-5G
SIAL259624-5GEA 38.4 EUR
SIAL259624-5G
4-Brombenzhydrazid, Sigma-Aldrich®
4-Brombenzhydrazid
Formel: C₇H₇BrN₂O
Molecular Weight: 215,05 g/mol
MDL: MFCD00007602
CAS-Nummer: 5933-32-4
EINECS: 227-681-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR