Dinatriumbenzol-1,3-disulfonat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1,3-Benzenedisulphonic acid disodium salt, Benzene-1,3-disulphonic acid disodium salt

M-Clarity™ Qualitätsstufe = MQ100

259802-100G
SIAL259802-100GEA 66 EUR
SIAL259802-100G
Dinatriumbenzol-1,3-disulfonat, Sigma-Aldrich®
Dinatriumbenzol-1,3-disulfonat
Formel: C₆H₄Na₂O₆S₂
Molecular Weight: 282,21 g/mol
MDL: MFCD00007492
CAS-Nummer: 831-59-4
EINECS: 212-606-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR