(±)-1,2-Decandiol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (±)-1,2-Dihydroxydecan

M-Clarity™ Qualitätsstufe = MQ100

260320-50G
SIAL260320-50GEA 103 EUR
SIAL260320-50G
(±)-1,2-Decandiol, Sigma-Aldrich®
(±)-1,2-Decandiol
Formel: C₁₀H₂₂O₂
Molecular Weight: 174,28 g/mol
MDL: MFCD00010739
CAS-Nummer: 1119-86-4
EINECS: 214-288-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR