N-Bromphthalimid, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

260908-25G
SIAL260908-25GEA 89.1 EUR
SIAL260908-25G
N-Bromphthalimid, Sigma-Aldrich®
N-Bromphthalimid
Formel: C₈H₄BrNO₂
Molecular Weight: 226,03 g/mol
MDL: MFCD00005888
CAS-Nummer: 2439-85-2
EINECS: 219-467-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR