1,1-Dimethylharnstoff, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

261394-25G
SIAL261394-25GEA 85.8 EUR
SIAL261394-25G
1,1-Dimethylharnstoff, Sigma-Aldrich®
1,1-Dimethylharnstoff
Formel: C₃H₈N₂O
MDL: MFCD00007959
CAS-Nummer: 598-94-7
EINECS: 209-957-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR