Methylcumalat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Methyl 2-oxo-2H-pyran-5-carboxylate

261432-10G
SIAL261432-10GEA 317 EUR
SIAL261432-10G
Methylcumalat, Sigma-Aldrich®
Methylcumalat
Formel: C₇H₆O₄
Molecular Weight: 154,12 g/mol
MDL: MFCD00010120
CAS-Nummer: 6018-41-3
EINECS: 227-871-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR