trans-4-Pentylcyclohexancarbonsäure, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

261602-5G
SIAL261602-5GEA 70.4 EUR
SIAL261602-5G
trans-4-Pentylcyclohexancarbonsäure, Sigma-Aldrich®
trans-4-Pentylcyclohexanecarboxylic acid
Formel: C₁₁H₂₀O₂
Molecular Weight: 198,30 g/mol
MDL: MFCD00009964
CAS-Nummer: 38289-29-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR