Bromcyan 5 mol/l in Acetonitril, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Cyanogenbromid

261610-100ML
SIAL261610-100MLEA 82.2 EUR
SIAL261610-100ML
Bromcyan 5 mol/l in Acetonitril, Sigma-Aldrich®
Bromcyan
Formel: CBrN
Molecular Weight: 105,92 g/mol
MDL: MFCD00011597
CAS-Nummer: 506-68-3
EINECS: 208-051-2
UN: 3286
ADR: 3,I

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR