Pentafluornitrobenzol, Sigma-Aldrich®

Lieferant: Merck
Achtung

Synonyme: 1,2,3,4,5-Pentafluoro-6-nitrobenzene

M-Clarity™ Qualitätsstufe = MQ100

262218-5G
SIAL262218-5GEA 125 EUR
SIAL262218-5G
Pentafluornitrobenzol, Sigma-Aldrich®
Pentafluornitrobenzol
Formel: C₆F₅NO₂
Molecular Weight: 213,06 g/mol
MDL: MFCD00007049
CAS-Nummer: 880-78-4
EINECS: 212-915-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR