Bis(cyclopentadienyl)ruthenium, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Ruthenocene, Bis(cyclopentadienyl)ruthenium(II)

M-Clarity™ Qualitätsstufe = MQ100

262455-1G
SIAL262455-1GEA 139 EUR
SIAL262455-1G
Bis(cyclopentadienyl)ruthenium, Sigma-Aldrich®
Bis(cyclopentadienyl)ruthenium
Formel: C₁₀Ru
Molecular Weight: 221,18 g/mol
MDL: MFCD00001442
CAS-Nummer: 1287-13-4
EINECS: 215-065-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR