3,5-Bis(trifluormethyl)phenylacetonitril, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-(3,5-Bis(trifluoromethyl)phenyl)acetonitrile

M-Clarity™ Qualitätsstufe = MQ100

263427-5G
SIAL263427-5GEA 81.8 EUR
SIAL263427-5G
3,5-Bis(trifluormethyl)phenylacetonitril, Sigma-Aldrich®
3,5-Bis(trifluormethyl)phenylacetonitril
Formel: C₁₀H₅F₆N
MDL: MFCD00009904
CAS-Nummer: 85068-32-2
EINECS: 285-294-0
UN: 3276
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR