2,3-Difluorbenzoesäure, Sigma-Aldrich®

Lieferant: Merck
Achtung


M-Clarity™ Qualitätsstufe = MQ100

263516-10G
SIAL263516-10GEA 52.8 EUR
SIAL263516-10G
2,3-Difluorbenzoesäure, Sigma-Aldrich®
2,3-Difluorobenzoic acid
Formel: C₇H₄F₂O₂
MDL: MFCD00010267
CAS-Nummer: 4519-39-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR