2-Mercapto-5-nitrobenzimidazol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-Mercapto-5-nitro-1H-benzimidazol, 5-Nitrobenzimidazol-2-thiol

M-Clarity™ Qualitätsstufe = MQ100

263575-5G
SIAL263575-5GEA 40.6 EUR
SIAL263575-5G
2-Mercapto-5-nitrobenzimidazol, Sigma-Aldrich®
2-Mercapto-5-nitrobenzimidazole
Formel: C₇H₅N₃O₂S
Molecular Weight: 195,2 g/mol
Schmelzpunkt: 285…287 °C
MDL: MFCD00804727
CAS-Nummer: 6325-91-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR