4-Iodbenzaldehyd, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

263826-1G
SIAL263826-1GEA 108 EUR
SIAL263826-1G
4-Iodbenzaldehyd, Sigma-Aldrich®
4-Iodobenzaldehyde
Formel: C₇H₅IO
Molecular Weight: 232,02 g/mol
MDL: MFCD00039576
CAS-Nummer: 15164-44-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR