Wolfram(IV)chlorid, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

263974-25G 263974-5G
SIAL263974-25GEA 387 EUR
SIAL263974-25G SIAL263974-5G
Wolfram(IV)chlorid, Sigma-Aldrich®
Wolfram(IV)chlorid
Formel: Cl₄W
MDL: MFCD00012027
CAS-Nummer: 13470-13-8
UN: 3260
ADR: 8,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR