Indium, powder, Sigma-Aldrich®
Lieferant: Merck
MQ100
264032-25G
264032-5G
SIAL264032-25GEA
340
EUR
SIAL264032-25G
SIAL264032-5G
Indium, powder, Sigma-Aldrich®
Indium
Formel:
In Molecular Weight: 114.82 g/mol Siedepunkt: 2000 Schmelzpunkt: 156 Dichte: 7.31 g/cm³ (20 °C) |
MDL:
MFCD00134048 CAS-Nummer: 7440-74-6 EINECS: 231-180-0 UN: 3089 ADR: 4.1 III Merck Index: 13,04971 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...