Chlor-tris-pyrrolidinophosphoniumhexafluorophosphat, Sigma-Aldrich®

Lieferant: Merck

26564-25G-F
SIAL26564-25G-FEA 313 EUR
SIAL26564-25G-F
Chlor-tris-pyrrolidinophosphoniumhexafluorophosphat, Sigma-Aldrich®
Chlor-tris-pyrrolidinophosphoniumhexafluorophosphat
Formel: C₁₂H₂₄ClF₆N₃P₂
MDL: MFCD00210035
CAS-Nummer: 133894-48-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR