Mangan, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

266167-500G
SIAL266167-500GEA 85.5 EUR
SIAL266167-500G
Mangan, Sigma-Aldrich®
Manganese
Formel: Mn
MDL: MFCD00011111
CAS-Nummer: 7439-96-5
EINECS: 231-105-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR