Chrom, Pulver, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

266299-100G 266299-500G
SIAL266299-100GEA 55 EUR
SIAL266299-100G SIAL266299-500G
Chrom, Pulver, Sigma-Aldrich®
Chromium
Formel: Cr
MDL: MFCD00010944
CAS-Nummer: 7440-47-3
EINECS: 231-157-5
UN: 3089
ADR: 4.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR