Kobalt, Pulver, Sigma-Aldrich®
Lieferant: Merck
|
Gefahr
|
M-Clarity™ Qualitätsstufe = MQ100
266647-250G
266647-50G
SIAL266647-250GEA
279
EUR
SIAL266647-250G
SIAL266647-50G
Kobalt, Pulver, Sigma-Aldrich®
Cobalt
Formel:
Co |
MDL:
MFCD00010935 CAS-Nummer: 7440-48-4 EINECS: 231-158-0 UN: 3089 ADR: 4.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...