Hafnium, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

266809-100G
SIAL266809-100GEA 572 EUR
SIAL266809-100G
Hafnium, Sigma-Aldrich®
Hafnium
Formel: Hf
MDL: MFCD00011032
CAS-Nummer: 7440-58-6
EINECS: 231-166-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR