2-Methyl-3-pyridinol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3-Hydroxy-2-methylpyridin

M-Clarity™ Qualitätsstufe = MQ100

268534-5G
SIAL268534-5GEA 75.3 EUR
SIAL268534-5G
2-Methyl-3-pyridinol, Sigma-Aldrich®
2-Methylpyridin-3-ol
Formel: C₆H₇NO
Molecular Weight: 109,13 g/mol
MDL: MFCD00082538
CAS-Nummer: 1121-25-1
EINECS: 214-327-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR