2,4-Dibrom-6-nitroanilin, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

268755-1G
SIAL268755-1GEA 45.8 EUR
SIAL268755-1G
2,4-Dibrom-6-nitroanilin, Sigma-Aldrich®
2,4-Dibrom-6-nitroanilin
Formel: C₆H₄Br₂N₂O₂
Molecular Weight: 295,92 g/mol
Schmelzpunkt: 127 °C
MDL: MFCD00012107
CAS-Nummer: 827-23-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR