L(-)-tert-Leucin (H-Tle-OH), Sigma-Aldrich®

Lieferant: Merck

Synonyme: (S)-2-Amino-3,3-dimethylbutanoic acid, H-Tle-OH

MQ200

269107-25G 269107-5G
SIAL269107-25GEA 173 EUR
SIAL269107-25G SIAL269107-5G
L(-)-tert-Leucin (H-Tle-OH), Sigma-Aldrich®
L(-)-tert-Leucin (H-Tle-OH)
Formel: C₆H₁₃NO₂
Molecular Weight: 131.2 g/mol
MDL: MFCD00064218
CAS-Nummer: 20859-02-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR