D-tert-Leucin (H-D-Tle-OH), Sigma-Aldrich®

Lieferant: Merck

Synonyme: H-D-Tle-OH

M-Clarity™ Qualitätsstufe = MQ100

269115-500MG
SIAL269115-500MGEA 215 EUR
SIAL269115-500MG
D-tert-Leucin (H-D-Tle-OH), Sigma-Aldrich®
D-tert-Leucin (H-D-Tle-OH)
Formel: C₆H₁₃NO₂
Molecular Weight: 131.2 g/mol
MDL: MFCD00004265
CAS-Nummer: 26782-71-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR