Ethyl-4,4,4-trifluorcrotonat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Ethyl (E)-4,4,4-trifluorobut-2-enoate, Ethyl 4,4,4-trifluorocrotonate

M-Clarity™ Qualitätsstufe = MQ100

269697-5G
SIAL269697-5GEA 72.8 EUR
SIAL269697-5G
Ethyl-4,4,4-trifluorcrotonat, Sigma-Aldrich®
Ethyl-4,4,4-trifluorcrotonat
Formel: C₆H₇F₃O₂
Molecular Weight: 168,12 g/mol
MDL: MFCD00009903
CAS-Nummer: 25597-16-4
UN: 3272
ADR: 3,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR