Di-tert-butyl-3-oxoglutarat, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ200

270814-5G
SIAL270814-5GEA 1220 EUR
SIAL270814-5G
Di-tert-butyl-3-oxoglutarat, Sigma-Aldrich®
Di-tert-butyl-3-oxoglutarat
Formel: C₁₃H₂₂O₅
Molecular Weight: 258,31 g/mol
CAS-Nummer: 28009-80-5
EINECS: 248-775-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR