Poly(vinyl sulphate) potassium salt, M.W. ~170,000, Pulver, Sigma-Aldrich®

Lieferant: Merck
Achtung

Synonyme: Potassium ethenyl sulphate, Potassium poly(vinyl sulphate), Polyvinyl sulphate potassium salt

M-Clarity™ Qualitätsstufe = MQ100

271969-1G 271969-5G
SIAL271969-1GEA 109 EUR
SIAL271969-1G SIAL271969-5G
Poly(vinyl sulphate) potassium salt, M.W. ~170,000, Pulver, Sigma-Aldrich®
Poly(vinyl sulphate) potassium salt
Formel: [CH₂CH(OSO₃K)]n
MDL: MFCD00081923
CAS-Nummer: 26182-60-5

Bestellen

Spezifikation Testergebnisse

Potassium (K) by ICP Analysis 22.0 - 27.0 %
Solubility (Turbidity) (C = 2% in H2O (Heat)) Clear to Slightly Hazy


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR